Iodacetamid, Millipore®

Lieferant: Merck Millipore Calbiochem
Achtung

Synonyme: Iodessigsäureamid

407710-25
407710-25EA 190 EUR
407710-25
Iodacetamid, Millipore®
Iodacetamid
Formel: ICH₂CONH₂
Molecular Weight: 184.96 g/mol
Siedepunkt: 266
Schmelzpunkt: 91…94 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008028
CAS-Nummer: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR