Graphit (natürlich) ≥99,9995% (Metall-Basis), Pulver -200 mesh
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40795.TB
40795.TC
40795.TD
40795.TBEA
188
EUR
40795.TB
40795.TC
40795.TD
Graphit (natürlich) ≥99,9995% (Metall-Basis), Pulver -200 mesh
Graphit
Formel:
C Molecular Weight: 12.01 g/mol Schmelzpunkt: 3827 Dichte: 2.09…2.23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144065 CAS-Nummer: 7782-42-5 EINECS: 231-955-3 UN: 1325 ADR: 4.1 III Merck Index: 12,04560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...