Ionomycin, Millipore®

Lieferant: Merck Millipore Calbiochem

407950-1 407950-10 407950-5
407950-1EA 230 EUR
407950-1 407950-10 407950-5
Ionomycin, Millipore®
Ionomycin
Formel: C₄₁H₇₂O₉
Molecular Weight: 709.02 g/mol
MDL: MFCD06798385
CAS-Nummer: 56092-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR