Graphit ≥99,9995% (Metall-Basis), Pulver -200 mesh

Lieferant: Thermo Fisher Scientific
Achtung

40797.TA 40797.TB 40797.TC 40797.TD
40797.TAEA 56.3 EUR
40797.TA 40797.TB 40797.TC 40797.TD
Graphit ≥99,9995% (Metall-Basis), Pulver -200 mesh
Graphit
Formel: C
Schmelzpunkt: 3827 °C
Dichte: 2,09…2.23 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1,III
Merck Index: 12,04560

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR