Graphit ≥99,9995% (Metall-Basis), Pulver -325 mesh
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40798.TA
40798.TB
40798.TC
40798.TD
40798.TAEA
62.3
EUR
40798.TA
40798.TB
40798.TC
40798.TD
Graphit ≥99,9995% (Metall-Basis), Pulver -325 mesh
Graphit
Formel:
C Schmelzpunkt: 3827 °C Dichte: 2,09…2.23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144065 CAS-Nummer: 7782-42-5 EINECS: 231-955-3 UN: 1325 ADR: 4.1,III Merck Index: 12,04560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...