Acrylamid 99,9, ultrapure_bioreagent Electrophoresis, purified, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propenamid, Acrylsäureamid, Propensäureamid, 2-Propenamid

4081-01EA 0 EUR
4081-01
Acrylamid 99,9, ultrapure_bioreagent Electrophoresis, purified, J.T.Baker®
Acrylamid
Formel: C₃H₅NO
Molecular Weight: 71.08 g/mol
Siedepunkt: 232
Schmelzpunkt: 84.5
Dichte: 1.122 g/cm³ (20 °C)
Flammpunkt: 138
Lagertemperatur: Kühlschrank
MDL: MFCD00008032
CAS-Nummer: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 99.9 %
Store between 18°C and 26°C. 6.0 - 7.0
Assay (CH₂:CHCONH₂) ≥ 99.9 %
pH of 10% Solution in 0.1M NaCl 6.0 - 7.0
Insoluble in H₂O ≤ 0.005 %
Insoluble in Methanol ≤ 0.005 %
Acrylic Acid (CH₂:CHCOOH) ≤ 0.001 %
Absorbance of a 1% Solution at 290 nm (1-cm path vs water)(au) ≤ 0.100
Conductivity of 35% Solution, µmho ≤ 2.5
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Heavy Metals (as Pb) ≤ 1 ppm
Iron (Fe) ≤ 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR