Eisen 99,95 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Lieferant: Thermo Fisher Scientific
40854.KS
40854.KSEA
219
EUR
40854.KS
Eisen 99,95 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Iron
Purity: 99.95%
CAS No.: 7439-89-6
MDL No.: MFCD00010999
Magnetron sputtering source
Formel:
Fe Molecular Weight: 55.85 g/mol Siedepunkt: 3000 Schmelzpunkt: 1535 Dichte: 7.86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010999 CAS-Nummer: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...