Mangan ≥99,95% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 6.35 mm (0.250 in)
Lieferant: Thermo Fisher Scientific
40863.KS
40863.KSEA
983
EUR
40863.KS
Mangan ≥99,95% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 6.35 mm (0.250 in)
Manganese
Purity: 99.95%
CAS No.: 7439-96-5
Physical Form: Hot pressed
Odour: Odourless
MDL No.: MFCD00011111
Formel:
Mn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011111 CAS-Nummer: 7439-96-5 EINECS: 231-105-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...