Ammoniumhydrogencarbonat ≥99% Eluentzusatz, für die LC-MS, Fluka™
Lieferant: Honeywell Chemicals
|
Achtung
|
40867-50G
40867-50GEA
50.96
EUR
40867-50G
Ammoniumhydrogencarbonat ≥99% Eluentzusatz, für die LC-MS, Fluka™
Ammoniumhydrogencarbonat
Formel:
NH₄HCO₃ Schmelzpunkt: 60 °C Dichte: 1,58 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012138 CAS-Nummer: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Spezifikation Testergebnisse
Assay | 99.5 - 101.0 % |
pH (1 M, 20°C) | 7.0 - 8.5 |
Solution 1 M in H₂O | complying |
Aluminium (Al) | Max. 5 ppm |
Barium (Ba) | Max. 5 ppm |
Bismuth (Bi) | Max. 5 ppm |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 50 ppm |
Lithium (Li) | Max. 5 ppm |
Magnesium (Mg) | Max. 500 ppm |
Manganese (Mn) | Max. 5 ppm |
Molybdenum (Mo) | Max. 5 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Strontium (Sr) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Chloride (Cl) | Max. 50 ppm |
Total S (as SO₄) | Max. 50 ppm |
Suitability for the LC-MS | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...