Nickel iron ≥99,95% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Scientific
Achtung

40886.KS
40886.KSEA 412 EUR
40886.KS
Nickel iron ≥99,95% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Nickel iron

Purity: 99.95% Molecular Formula: Ni:Fe; 81:19 wt% MDL No.: MFCD01741928


Applications: Magnetron sputtering source

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR