Hastelloy C® Legierung, Drahtgeflecht 20 mesh woven from 0.23mm (0.009in)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40938.FL
40938.GT
40938.HB
40938.FLEA
38.2
EUR
40938.FL
40938.GT
40938.HB
Hastelloy C® Legierung, Drahtgeflecht 20 mesh woven from 0.23mm (0.009in)
Hastelloy C® alloy
Physical Form: Wire Cloth
MDL No.: MFCD00801065
Applications: High strength, corrosion resistant alloy
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...