Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtform, Ø 0.127 mm (0.005 in) Type 304
Lieferant: Thermo Scientific
|
Gefahr
|
40944.BZ
40944.BZEA
54.1
EUR
40944.BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtform, Ø 0.127 mm (0.005 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odour: Odourless
MDL No.: MFCD00148499
Applications: Filtration, electrodes
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...