Diethylether 99+% inhibitorfrei für die Spektrophotometrie hemmstofffrei
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 60-29-7
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: -45°(-49°F)
Physical Form: Liquid
Density: 0.713
Refractive Index: 1.3525
Appearance: Colourless
Odour: Sweet
Melting Point: -116°
Boiling Point: 34.6°
Merck Reference: 14,3806
UN No.: 1155
MDL No.: MFCD00011646
Solvent for U.V. spectrophotometric and general use
Formel:
(CH₃CH₂)₂O Siedepunkt: 34,6 °C Schmelzpunkt: –116 °C Dichte: 0,713 Flammpunkt: –45 °C (–49 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011646 CAS-Nummer: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 14,3806 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...