(Triphenylphosphin)gold(I)bromid ≥99,99% (Metall-Basis), Premion®
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Bromo(triphenylphosphine)gold(I)
41002.03
41002.03EA
259
EUR
41002.03
(Triphenylphosphin)gold(I)bromid ≥99,99% (Metall-Basis), Premion®
(Triphenylphosphine)gold(I) bromide
Purity: 99.99%
CAS No.: 14243-65-3
Molecular Formula: AuBr[P(C6H5)3]
Formula Weight: 539.17
Physical Form: Powder
Formel:
C₁₈H₁₅AuBrP Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049267 CAS-Nummer: 14243-65-3 EINECS: 238-118-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...