L(+)-Weinsäure, Granulat NF, FCC, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: (R,R)-(+)-2,3-Dihydroxybutandisäure, (R,R)-(+)-2,3-Dihydroxy-1,4-butandisäure, L(+)-Threarsäure, natürliche Weinsäure, (R,R)-(+)-2,3-Dihydroxysuccinsäure, (R,R)-(+)-Weinsäure, (R,R)-(+)-2,3-Dihydroxybernsteinsäure

4104-01EA 205.76 EUR
4104-01
L(+)-Weinsäure, Granulat NF, FCC, J.T.Baker®
L(+)-Weinsäure
Formel: C₄H₆O₆
Molecular Weight: 150,09 g/mol
Siedepunkt: 399 °C (1013 hPa)
Schmelzpunkt: 168…170 °C
Dichte: 1,759 g/cm³ (20 °C)
Flammpunkt: 150 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064207
CAS-Nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product
GMP Manufactured Product
Meets N.F. Requirements
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+) 12.0 - 13.0 Degree
NF - Loss on Drying ≤0.5 %
NF - Residue on Ignition ≤0.1 %
NF - Oxalate (C₂O₄) Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Assay (HOCO(CHOH)₂COOH) (dried basis) 99.7 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+) 12.0 - 13.0 Degree
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.5 %
FCC - Residue on Ignition ≤0.1 %
FCC - Oxalate (C₂O₄) Passes Test
FCC - Sulfate (SO₄) Passes Test
FCC - Assay (HOCO(CHOH)₂COOH) (dried basis) 99.7 - 100.5 %
Manufacturer Verification Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR