Zirkonium(IV)sulfat Tetrahydrat ≥98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

41041.22 41041.36 41041.A3
41041.22EA 46.5 EUR
41041.22 41041.36 41041.A3
Zirkonium(IV)sulfat Tetrahydrat ≥98% (Metall-Basis)
Zirkonium(IV)sulfat Tetrahydrat

Purity: 98+% CAS No.: 7446-31-3 Molecular Formula: Zr(SO4)2.4H2O Formula Weight: 355.40 (283.34anhy) Physical Form: -20 Mesh Density: 3.22 Appearance: White Solubility: Soluble in water Merck Reference: 14,10182 MDL No.: MFCD00149267


Catalyst support

Formel: O₈S₂Zr·4H₂O
Dichte: 3,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00149267
CAS-Nummer: 7446-31-3
EINECS: 238-694-4
Merck Index: 14,10182

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR