TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid) ≥99,0% Made from Tromethamine USP, Biotechreagenz, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Tris(hydroxymethyl)methylamin Hydrochlorid, 2-Amino-2-(hydroxymethyl)propan-1,3-diolhydrochlorid, 2-Amino-2-(hydroxymethyl)-1,3-propandiol HCl, Tris(hydroxymethyl)methylammoniumchlorid, TRIS HCl, THAM HCl, Trometamol Hydrochlorid

4106-01EA 349.85 EUR
4106-01 4106-05 4106-07 4106-09 4106-F9 4106-F7
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid) ≥99,0% Made from Tromethamine USP, Biotechreagenz, J.T.Baker®
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)

GMP manufactured product. The infrared spectrum conforms to the standard.

Formel: NH₂C(CH₂OH)₃·HCl
Molecular Weight: 157.6 g/mol
Schmelzpunkt: 147…153 °C
Dichte: 1.05 g/ml (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012590
CAS-Nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product ≥99.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.06
Assay (NH₂C(CH₂OH)₃·HCl) ≥99.0 %
Appearance (colorless crystals to white crystalline powder) Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Identification (by IR) Passes Test
Loss on Drying at 105°C ≤0.5 %
Endotoxin Concentration (EU/g) ≤2.5
Melting Point 147 - 153 °C
pH of 0.5M Solution at 25°C 3.5 - 5.0
Trace Impurities - Arsenic (As) ≤10 ppm
Trace Impurities - Calcium (Ca) ≤10.0 ppm
Trace Impurities - Copper (Cu) ≤10 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Trace Impurities - Lead (Pb) ≤10 ppm
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR