Silicon(IV) nitride, MgO binder, sputtering target ≥99,9% (Metall-Basis), Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Scientific

41088.KS
41088.KSEA 979 EUR
41088.KS
Silicon(IV) nitride, MgO binder, sputtering target ≥99,9% (Metall-Basis), Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Silicon(IV) nitride, MgO binder, sputtering target

Purity: 99.9% Molecular Formula: Si3N4 Formula Weight: 140.28 MDL No.: MFCD00011230


Applications: Magnetron sputtering source

Formel: Si₃N₄
Molecular Weight: 140,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011230
EINECS: 234-796-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR