Silicon(IV) nitride, MgO binder, sputtering target ≥99,9% (Metall-Basis), Ø 76.2 mm (3.0 in), Dicke 3.18 mm (0.125 in)
Lieferant: Thermo Scientific
41090.KS
41090.KSEA
1358
EUR
41090.KS
Silicon(IV) nitride, MgO binder, sputtering target ≥99,9% (Metall-Basis), Ø 76.2 mm (3.0 in), Dicke 3.18 mm (0.125 in)
Silicon(IV) nitride, MgO binder, sputtering target
Purity: 99.9%
Molecular Formula: Si3N4
Formula Weight: 140.28
MDL No.: MFCD00011230
Formel:
Si₃N₄ Molecular Weight: 140,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011230 EINECS: 234-796-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...