Titannitrid ≥99,5% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Fisher Scientific

41110.KS
41110.KSEA 821 EUR
41110.KS
Titannitrid ≥99,5% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Titannitrid

Purity: 99.5% CAS No.: 25583-20-4 Molecular Formula: TiN Formula Weight: 61.91 Physical Form: Hot pressed material Density: 5.22 Appearance: Gray Melting Point: 2950° MDL No.: MFCD00049596


Magnetron sputtering source

Formel: NTi
Schmelzpunkt: 2950 °C
Dichte: 5,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00049596
CAS-Nummer: 25583-20-4
EINECS: 247-117-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR