Zinkoxid ≥99,9% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Zink(II)oxid

41123.KS
41123.KSEA 868 EUR
41123.KS
Zinkoxid ≥99,9% (Metall-Basis), Aufdampfplatte, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Zinkoxid

Purity: 99.9% CAS No.: 1314-13-2 Molecular Formula: OZn Formula Weight: 81.37 Physical Form: Target Density: 5.606 Appearance: White to pale yellow Odour: Odourless Melting Point: 1975° UN No.: 3077 MDL No.: MFCD00011300


Magnetron sputtering source

Formel: ZnO
Molecular Weight: 81.39 g/mol
Dichte: 5.606
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR