Saccharin, Supelco®

Lieferant: Merck

41131-5G
41131-5GEA 53.8 EUR
41131-5G
Saccharin, Supelco®
Saccharin
Formel: C₇H₅NO₃S
MDL: MFCD00005866
CAS-Nummer: 81-07-2
EINECS: 201-321-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1819cd33305869e78f489fede76da9373f296a1&vid=a1819cd33305869e78f489fede76da9373f296a1&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR