Mörsermühle, RM 200
Lieferant: Retsch GmbH
20.455.0001
412-2909EA 5980 EUR
412-2909
Mörsermühle, RM 200
Mühlen
The mortar grinder RM 200 comminutes inorganic and organic substances to analytical fineness. It can mix and homogenise powders, suspensions and pastes. The grinder is also ideal for the uniform trituration of homeopathic and pharmaceutical products. Depending on the sample, amounts from approx. 10 to 190 ml can be processed in the RM 200.
  • Dry, wet and cryogenic grinding
  • Reproducible results due to adjustable pestle and scraper pressure
  • Easy exchange of mortar and pestle without tools
  • Closed, dust tight grinding chamber with windows
  • Wide range of materials for contamination-free grinding
* Depending on feed material and instrument configurations/settings
Bestellinformation: Pistillen und Mörser bitte separat bestellen.
Bitte beachten * Abhängig von Probenmaterial und Instrumentenkonfiguration/-einstellungen