Ball mill, vibratory micro mill PULVERISETTE 0
Lieferant: FRITSCH
00.6020.00
412-4181EA 2804 EUR
412-4181
Ball mill, vibratory micro mill PULVERISETTE 0
Mühlen
Micro vibratory mill - single ball- for fine comminution of dry laboratory samples or solids in suspension.
  • Effective comminution in a narrow, homogeneous particle size range
  • Loss-free grinding in a closed vessel – dry or in suspension
  • Cryogenic grinding and simple embrittling in the cryo-box
  • Adjustable oscillation amplitude for easy adaption of the vibration energy to the grinding sample
  • Digital timer
Bestellinformation: Standard equipment includes grinding head. Mortar and grinding ball have to ordered separately