Mortar grinder, PULVERISETTE 2
Lieferant: FRITSCH
02.2000.00
412-4183EA 5901 EUR
412-4183
Mortar grinder, PULVERISETTE 2
Mühlen
This mortar grinder is an ideal universal grinder in the laboratory for analysis, quality control and materials testing as well as for the mixing and homogenisation of pastes and creams on a laboratory scale. Ideal for wet and dry grinding, mixing and homogenisation.
  • For hard, medium-hard, soft, brittle, fibrous and abrasive materials
  • Particularly gentle grinding with no thermal load
  • Loss-free grinding due to closed grinding chamber
  • Cryogenic grinding with automatic timer for reproducible grinding times
Using liquid nitrogen it also grinds difficult-to-mill samples with moist, fibrous or elastic structures. Precisely adjustable pestle pressure and optimum scraper kinematics. Illuminated grinding chamber for optimum control.