Planetary mono mill, PULVERISETTE 6 classic line
Lieferant: FRITSCH
06.2000.00
412-4185EA 7764 EUR
412-4185
Planetary mono mill, PULVERISETTE 6 classic line
Mühlen
A planetary ball mill for the finest rapid, batch-wise comminution of hard to soft grinding material, dry or in suspension, down to colloidal fineness. Can also be used for mixing and homogenising of emulsions.
  • Large rotational speed range with accurate display of grinding parameters
  • Grinding chamber completely encapsulated but easy to open
  • Cooling of the grinding chamber with a built-in fan for long grinding times
  • Exactly reproducible grinding results thanks to a regulated drive, precise transmission ratio (toothed belt), programmable microprocessor control electronics
  • Smaller grinding bowls are also possible with an adapter
  • Splash-proof IP65 membrane keyboard and easy cleaning of the grinding elements
Standards: Splash-proof IP 65 membrane keyboard and easy cleaning of the grinding elements.
Bestellinformation: Grinding bowls and grinding balls are not included in the standard instrument package and must be ordered separately.