Planetary micro mill, PULVERISETTE 7 classic line
Lieferant: FRITSCH
07.4000.00
412-4201EA 8308 EUR
412-4201
Planetary micro mill, PULVERISETTE 7 classic line
Mühlen
Planetary micro mill for the uniform, and extremely fine comminution of very small samples of hard to soft grinding material, dry or in suspension, down to colloidal fineness. For mixing and perfect homogenising of emulsions or pastes. The grinding sample is comminuted by high-energy impacts from grinding balls and friction between balls and the grinding bowl wall.
  • Precise rotational speed regulation with display of set/actual values and reversing function
  • Programme timer for grinding operation and cooling phases
  • Grinding chamber completely encapsulated but easy to open
  • Cooling of the grinding chamber with a built-in fan for long grinding times
  • Ergonomic IP 64 membrane keyboard
  • Maintenance-free drive with asynchronous motor and frequency converter