Disk mill, PULVERISETTE 13
Lieferant: FRITSCH
13.1030.00
412-4301EA 17099 EUR
412-4301
Disk mill, PULVERISETTE 13
Mühlen
Disc mill for batch-wise or continuous fine grinding of hard-brittle to medium-hard solids.
  • High throughput up to 150 kg/h
  • Fine grinding down to 0,1 mm
  • Simple operation with fast, easy cleaning
  • Fast grinding disc change
  • Precision setting of the final fineness even during grinding
The material to be ground is comminuted by pressure and shearing action between two counteracting grinding disks with coarse interlocking teeth. The ground material automatically falls down through an pre-set grinding gap into the collection drawer. The desired final fineness can be defi ned from the outside by changing the gap width.
Bestellinformation: Grinding discs are offered in 4 different materials and must be ordered separately fromthe mill.