Boats, platinum
Lieferant: Thermo Fisher Scientific
41216.KS
41216.KSEA 330 EUR
41216.KS
Boats, platinum
Schiffchen Verbrennungsschiffchen
Combustion boats, Platinum.
  • High melting point
  • High resistance against corrosion
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum boats have become essential for the preparation of samples in analytical laboratories.