Cobalt(II)-2-methoxyethoxide 5% (w/v) in Methylglycol , 99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 142600-61-1
Molecular Formula: C6H14CoO4
Formula Weight: 209.11
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Dark brown
Boiling Point: 124°
UN No.: UN1188
MDL No.: MFCD00798554
Formel:
C₆H₁₄CoO₄ Siedepunkt: 124 °C Dichte: 0,96 Flammpunkt: 40 °C (104 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798554 CAS-Nummer: 142600-61-1 UN: 1188 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...