4-(2-Hydroxyethyl)benzonitril 98%

Lieferant: Thermo Fisher Scientific
Gefahr

41283.03 41283.06
41283.03EA 262 EUR
41283.03 41283.06
4-(2-Hydroxyethyl)benzonitril 98%
4-(2-Hydroxyethyl)benzonitril

Purity: 98% CAS No.: 69395-13-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Physical Form: Powder/Chunk Boiling Point: 116-118°/0.5mm UN No.: 3439 MDL No.: MFCD00052461


Organic intermediate

Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Siedepunkt: 116…118 °C (0.5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052461
CAS-Nummer: 69395-13-7
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR