Mucicarmin-Lösung (Mayer) für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

41325-100ML-F
41325-100ML-FEA 63.4 EUR
41325-100ML-F
Mucicarmin-Lösung (Mayer) für die Mikroskopie, Sigma-Aldrich®
Mucicarmin

Most mucins contain serveral glycoproteins and can therefore be detected by stains for carbohydrates. Best carmine stain is specifically used for glycogens. Mucicarmine is an empirical stain. When applied for fungal staining, it stains the spores and hyphae as pale to dark red.

Lagertemperatur: Raumtemperatur
MDL: MFCD00071289
CAS-Nummer: 51395-97-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR