Xylenolorange Tetranatriumsalz ACS
Lieferant: Thermo Fisher Scientific
41379.04
41379.09
41379.18
41379.04EA
70.4
EUR
41379.04
41379.09
41379.18
Xylenolorange Tetranatriumsalz ACS
Xylenolorange Tetranatriumsalz
CAS No.: 3618-43-7
Molecular Formula: C31H28N2Na4O13S
Formula Weight: 760.59
Physical Form: Powder
Odour: Odourless
Melting Point: 210° dec.
MDL No.: MFCD00044293
Formel:
C₃₁H₂₈N₂Na₄O₁₃S Schmelzpunkt: 210 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044293 CAS-Nummer: 3618-43-7 EINECS: 222-805-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...