3-Nitrophthalimid 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

414034-10G
414034-10GEA 238 EUR
414034-10G
3-Nitrophthalimid 97%, Sigma-Aldrich®
3-Nitrophthalimid
Formel: C₈H₄N₂O₄
Molecular Weight: 192,13 g/mol
Schmelzpunkt: 218 °C
Lagertemperatur: Room Temperature
MDL: MFCD00041852
CAS-Nummer: 603-62-3
EINECS: 210-051-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b7bb45c2bc0a4e193d7f009886d743974ef41fe&vid=a066a99d3a38c07cf42aeb8e61e2c0293b501a69&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Conforms to Requirements
Faint to Dark Yellow
Appearance (Form) Conforms to Requirements
Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon 48.2 - 51.8%
Nitrogen 14.0 - 15.1%
Purity (HPLC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR