Methanol ≥99,9%, CHROMASOLV™ zur GC/MS Analyse volatiler organischer Stoffe, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
414816-1L
414816-1LEA
149.04
EUR
414816-1L
Methanol ≥99,9%, CHROMASOLV™ zur GC/MS Analyse volatiler organischer Stoffe, Riedel-de Haën™
Methanol
Honeywell Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
Beilstein Number: 1098229
Formel:
CH₃OH Siedepunkt: 64…65 °C (1,013 hPa) Schmelzpunkt: –98 °C Dichte: 0,79 g/cm³ (20 °C) Flammpunkt: 11 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004595 CAS-Nummer: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Spezifikation Testergebnisse
Autoignition Temperature | 725 °F |
IR spectrum | complying |
Assay (GC) | Min. 99.90 % |
Non-volatile matter | Max. 1 ppm |
Water (Karl Fischer) | Max. 0.2 % |
Impurity Profile | complying |
Vapor Pressure | 97.68 mmHg ( 20 °C) |
Refractive Index | n20/D 1.329 (lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...