Molybdenum(V) isopropoxide ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

41484.04 41484.02
41484.04EA 319 EUR
41484.04 41484.02
Molybdenum(V) isopropoxide ≥99% (Metall-Basis)
Molybdenum(V) isopropoxide

Purity: 99+% CAS No.: 209733-38-0 Molecular Formula: C15H35MoO5 Formula Weight: 391.04 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Yellow to brown to black UN No.: UN3181 MDL No.: MFCD00210636


Applications: Precursor used in sol-gel process

Formel: C₁₅H₃₅MoO₅
Molecular Weight: 391.39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210636
CAS-Nummer: 209733-38-0
UN: 3181
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR