Molybdenum(V) isopropoxide ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99+%
CAS No.: 209733-38-0
Molecular Formula: C15H35MoO5
Formula Weight: 391.04
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown to black
UN No.: UN3181
MDL No.: MFCD00210636
Applications: Precursor used in sol-gel process
Formel:
C₁₅H₃₅MoO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210636 CAS-Nummer: 209733-38-0 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...