Cobalt(II)-phthalocyanin

Lieferant: Thermo Fisher Scientific
Achtung

41496.03 41496.06 41496.14
41496.03EA 36.4 EUR
41496.03 41496.06 41496.14
Cobalt(II)-phthalocyanin
Cobalt(II) Phthalocyanine

CAS No.: 3317-67-7 Molecular Formula: C32H16N8Co Formula Weight: 571.47 Physical Form: Powder Appearance: Dark blue to purple Odour: Odourless MDL No.: MFCD00010718

Formel: C₃₂H₁₆CoN₈
Lagertemperatur: Raumtemperatur
MDL: MFCD00010718
CAS-Nummer: 3317-67-7
EINECS: 222-012-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR