Lactophenol-picric acid-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Picric acid-lactophenol solution
41506-100ML-F
41506-100ML-FEA
44.3
EUR
41506-100ML-F
Lactophenol-picric acid-Lösung, Sigma-Aldrich®
Lactophenol-picric acid
Picric acid (2,4,6-trinitrophenol) has antibacterial as well as antifungal properties. It stains the fungal spores and hyphae a pale to dark yellow color.
Lagertemperatur:
Raumtemperatur |
ADR:
8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...