Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex-Lösung ≥98%
Lieferant: Thermo Fisher Scientific
CAS No.: 68478-92-2
Molecular Formula: (C8H18OSi2)1.5Pt
Formula Weight: 474.68
Flash Point:
Applications: Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
Formel:
C₈H₁₈OPtSi₂ Dichte: 0,984 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151662 CAS-Nummer: 68478-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...