Sulfanilsäure 98,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98.0-102.0%
CAS No.: 121-57-3
Molecular Formula: C6H7NO3S
Formula Weight: 173.19
Physical Form: Powder
Melting Point: ca 365°
Beilstein Reference: 908765
Merck Reference: 14,8926
MDL No.: MFCD00007886
Reagent for chromatographic detection of histidine
Formel:
4-(H₂N)C₆H₄SO₃H Siedepunkt: 363 °C (1013 hPa) Schmelzpunkt: ∼365 °C Dichte: 1,485 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007886 CAS-Nummer: 121-57-3 EINECS: 204-482-5 Merck Index: 14,8926 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...