Antimon(III)bromid 99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 7789-61-9
Molecular Formula: SbBr3
Formula Weight: 361.48
Physical Form: Powder
Density: 4.148
Refractive Index: 1.74
Sensitivity: Moisture Sensitive
Odour: Characteristic
Melting Point: 96.6°
Boiling Point: 280°
Merck Reference: 14,706
UN No.: 3260
MDL No.: MFCD00016317
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...