Cadmiumacetat Dihydrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

41581.22 41581.36 41581.A3
41581.22EA 57.7 EUR
41581.22 41581.36 41581.A3
Cadmiumacetat Dihydrat ≥98%
Cadmiumacetat Dihydrat

Purity: 98% CAS No.: 5743-04-4 Molecular Formula: Cd(OOCCH3)2.2H2O Formula Weight: 266.54 (230.50anhy) Physical Form: Powder Density: 2.01 Appearance: White Solubility: Freely soluble in water, soluble in alcohol Merck Reference: 14,1614 UN No.: 2570 MDL No.: MFCD00012449


In cadium electroplating

Formel: (H₃CCOO)₂Cd·2H₂O
Siedepunkt: 105…107 °C (4 mmHg)
Schmelzpunkt: 256 °C
Dichte: 2,01
Lagertemperatur: Raumtemperatur
MDL: MFCD00150020
CAS-Nummer: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 14,1614

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR