Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Folie, Dicke 0.25 mm (0.01 in) Type 304
Lieferant: Thermo Scientific
41584.GH
41584.GHEA
70.1
EUR
41584.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Folie, Dicke 0.25 mm (0.01 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odour: Odourless
MDL No.: MFCD00148499