tri-Kaliumorthophosphat Monohydrat ≥95%

Lieferant: Thermo Fisher Scientific
Achtung

41586.36 41586.A3
41586.36EA 117 EUR
41586.36 41586.A3
tri-Kaliumorthophosphat Monohydrat ≥95%
tri-Kaliumorthophosphat Monohydrat

Purity: 95% CAS No.: 27176-10-9 Molecular Formula: K3PO4.H2O Formula Weight: 230.28 (212.27anhy) Physical Form: Granular Density: 2.564 MDL No.: MFCD00150410

Formel: K₃O₄P·1H₂O
Dichte: 2,564
Lagertemperatur: Raumtemperatur
MDL: MFCD00150410
CAS-Nummer: 27176-10-9
EINECS: 231-907-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR