Copper Standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
41621-100ML
41621-100MLEA
271
EUR
41621-100ML
Copper Standard for ICP-MS, TraceCERT®, Supelco®
Standards
Kationischer Standards
Kupfer-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034