Platin-Rhodium-Legierung (80:20 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.1 mm (0.004 in)
Lieferant: Thermo Scientific
41623.FF
41623.FFEA
2629
EUR
41623.FF
Platin-Rhodium-Legierung (80:20 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.1 mm (0.004 in)
Platin-Rhodium-Legierung (80:20 gew.%)
Purity: 99.9%
Molecular Formula: Pt:Rh; 80:20 wt%
Physical Form: ≈1.17g/25x25mm
Odour: Odourless
MDL No.: MFCD00198186
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...