Platin ≥99,95% (Metall-Basis), dünne Folie, hart, Dicke 0.00125 mm (0.00005 in)
Lieferant: Thermo Fisher Scientific
41632.FF
41632.FFEA
1468
EUR
41632.FF
Platin ≥99,95% (Metall-Basis), dünne Folie, hart, Dicke 0.00125 mm (0.00005 in)
Platinum
Purity: 99.95%
CAS No.: 7440-06-4
Molecular Formula: Not certified pinhole free
Physical Form: ≈0.017g/25x25mm, Temper: as rolled
Odour: Odourless
Formel:
Pt Siedepunkt: 3827 °C (1013 hPa) Schmelzpunkt: 1769 °C Dichte: 21,447 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011179 CAS-Nummer: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...