Bariumtitanat ≥99% (Metall-Basis), große Fläche

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bariumtitanat, Bariumtitan(IV)oxid, Bariumtitanoxid, Bariumorthotitanat, Bariumtitanat(IV)

41633.14 41633.22
41633.14EA 81 EUR
41633.14 41633.22
Bariumtitanat ≥99% (Metall-Basis), große Fläche
Bariumtitanat

Purity: 99+% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 0.05-0.07 Micron APS Powder, S.A. 10-30m2/g Density: 5.85 Appearance: White to grey Odour: Odourless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Formel: BaO₃Ti
Molecular Weight: 233.21 g/mol
Schmelzpunkt: 1620 °C
Dichte: 5.85
Lagertemperatur: Raumtemperatur
MDL: MFCD00003447
CAS-Nummer: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,1000

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR