Lanthan(III)oxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.9%
CAS No.: 1312-81-8
Molecular Formula: La2O3
Formula Weight: 325.82
Physical Form: 3-12mm Sintered Lump
Density: 6.51
Melting Point: 2307°
Boiling Point: 4200°
Solubility: Insoluble in water. Soluble in dilute mineral acids
Merck Reference: 14,5363
MDL No.: MFCD00011071
For vacuum deposition
Formel:
La₂O₃ Siedepunkt: 4200 °C Schmelzpunkt: 2307 °C Dichte: 6,51 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011071 CAS-Nummer: 1312-81-8 EINECS: 215-200-5 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...